3-Aminoisonicotinsäure

Lieferant: Thermo Scientific

Synonyme: 3-Aminoisonicotinic Acid, 3-Aminopyridine-4-carboxylic acid

SP00054.250 SP00054DA
MAYBSP00054.250EA 0 EUR
MAYBSP00054.250 MAYBSP00054DA
3-Aminoisonicotinsäure
3-Aminoisonicotinsäure
Formel: C₆H₆N₂O₂
Molecular Weight: 138,13 g/mol
MDL: MFCD00137840
CAS-Nummer: 7579-20-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR