Ethyl-6-chlornicotinat
Lieferant: Thermo Scientific
Synonyme:
Ethyl 6-chloronicotinate
SPB05346.1
SPB05346.10
SPB05346.50
MAYBSPB05346.1EA
0
EUR
MAYBSPB05346.1
MAYBSPB05346.10
MAYBSPB05346.50
Ethyl-6-chlornicotinat
Ethyl-6-chlornicotinat
Formel:
C₈H₈ClNO₂ Molecular Weight: 185,61 g/mol |
MDL:
MFCD00082739 CAS-Nummer: 49608-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...