5-Brom-2-hydroxy-3-nitropyridin

Lieferant: Thermo Scientific

Synonyme: 5-Bromo-3-nitro-2-pyridinol, 5-Bromo-3-nitropyridin-2(1H)-one

SPB06123.1 SPB06123EA
MAYBSPB06123.1EA 0 EUR
MAYBSPB06123.1 MAYBSPB06123EA
5-Brom-2-hydroxy-3-nitropyridin
5-Brom-2-hydroxy-3-nitropyridin
Formel: C₅H₃BrN₂O₃
Molecular Weight: 218.99 g/mol
MDL: MFCD00023473
CAS-Nummer: 15862-34-7
EINECS: 239-989-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR