5-Chlor-2-hydroxy-3-nitropyridin

Lieferant: Thermo Scientific

Synonyme: 5-Chloro-3-nitropyridin-2-ol

SPB06124.1 SPB06124.10
MAYBSPB06124.1EA 0 EUR
MAYBSPB06124.1 MAYBSPB06124.10
5-Chlor-2-hydroxy-3-nitropyridin
5-Chlor-2-hydroxy-3-nitropyridin
Formel: C₅H₃ClN₂O₃
Molecular Weight: 174,54 g/mol
MDL: MFCD00114884
CAS-Nummer: 21427-61-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR