5-Chlor-2-fluor-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Chloro-2-fluoropyridine-3-boronic acid, (5-Chloro-2-fluoropyridin-3-yl)boronic acid, 5-Chloro-2-fluoro-3-pyridylboronic acid

BB-3818-1G BB-3818-5G
COBBBB-3818-1GEA 115 EUR
COBBBB-3818-1G COBBBB-3818-5G
5-Chlor-2-fluor-3-pyridinboronsäure
5-Chloro-2-fluoro-3-pyridineboronic acid
Formel: C₅H₄BClFNO₂
Molecular Weight: 175.35 g/mol
MDL: MFCD04972408
CAS-Nummer: 937595-70-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR