2,3-Dichlor-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2,3-Dichloropyridine-4-boronic acid, 2,3-Dichloro-4-pyridylboronic acid

BB-3926-5G BB-3926-1G BB-3926-25G
COBBBB-3926-5GEA 208 EUR
COBBBB-3926-5G COBBBB-3926-1G COBBBB-3926-25G
2,3-Dichlor-4-pyridinboronsäure
2,3-Dichloro-4-pyridineboronic acid
Formel: C₅H₄BCl₂NO₂
Molecular Weight: 191,81 g/mol
MDL: MFCD04038756
CAS-Nummer: 951677-39-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR