5-Chlor-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Chloropyridine-3-boronic acid, (5-Chloropyridin-3-yl)boronic acid

BB-5001-5G BB-5001-1G
COBBBB-5001-5GEA 249 EUR
COBBBB-5001-5G COBBBB-5001-1G
5-Chlor-3-pyridinboronsäure
5-Chloro-3-pyridineboronic acid
Formel: C₅H₅BClNO₂
Molecular Weight: 157,36 g/mol
MDL: MFCD06798243
CAS-Nummer: 872041-85-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR