2-Chlor-4-methoxy-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-5190-1G BB-5190-250M
COBBBB-5190-1GEA 1150 EUR
COBBBB-5190-1G COBBBB-5190-250M
2-Chlor-4-methoxy-5-pyridinboronsäure
2-Chlor-4-methoxy-5-pyridinboronsäure
Formel: C₆H₇BClNO₃
MDL: MFCD10699625
CAS-Nummer: 1072946-20-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR