2-Chlor-6-methoxy-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-6-methoxypyridine-3-lboronic acid

BB-5203-5G BB-5203-1G
COBBBB-5203-5GEA 1810 EUR
COBBBB-5203-5G COBBBB-5203-1G
2-Chlor-6-methoxy-3-pyridinboronsäure
2-Chloro-6-methoxy-3-pyridineboronic acid
Formel: C₆H₇BClNO₃
Molecular Weight: 187.39 g/mol
MDL: MFCD11053826
CAS-Nummer: 1072946-25-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR