2,5-Dichlor-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2,5-Dichloropyridine-4-boronic acid, 2,5-Dichloro-4-pyridylboronic acid

BB-5232-5G BB-5232-1G
COBBBB-5232-5GEA 232 EUR
COBBBB-5232-5G COBBBB-5232-1G
2,5-Dichlor-4-pyridinboronsäure
2,5-Dichloro-4-pyridineboronic acid
Formel: C₅H₄BCl₂NO₂
Molecular Weight: 191,81 g/mol
MDL: MFCD06798254
CAS-Nummer: 847664-64-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR