2-Chlor-3-fluor-5-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (6-Chloro-5-fluoropyridin-3-yl)boronic acid

BB-5290-1G BB-5290-250M
COBBBB-5290-1GEA 384 EUR
COBBBB-5290-1G COBBBB-5290-250M
2-Chlor-3-fluor-5-pyridinboronsäure
2-Chlor-3-fluor-5-pyridinboronsäure
Formel: C₅H₄BClFNO₂
Molecular Weight: 175,35 g/mol
MDL: MFCD04112500
CAS-Nummer: 1072946-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR