6-Chlor-2-fluor-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (6-Chloro-2-fluoropyridin-3-yl)boronic acid, 6-Chloro-2-methylpyridine-3-boronic acid, 6-Chloro-2-fluoropyridine-3-boronic acid

BB-5458-5G BB-5458-1G
COBBBB-5458-5GEA 1420 EUR
COBBBB-5458-5G COBBBB-5458-1G
6-Chlor-2-fluor-3-pyridinboronsäure
6-Chloro-2-fluoro-3-pyridineboronic acid
Formel: C₅H₄BClFNO₂
Molecular Weight: 175.35 g/mol
MDL: MFCD11504861
CAS-Nummer: 1256345-66-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR