2-Chlor-3-fluor-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-3-Fluoropyridine-4-Boronic Acid

BB-8244-5G BB-8244-1G
COBBBB-8244-5GEA 102 EUR
COBBBB-8244-5G COBBBB-8244-1G
2-Chlor-3-fluor-4-pyridinboronsäure
2-Chloro-3-fluoro-4-pyridineboronic acid
Formel: C₅H₄BClFNO₂
Molecular Weight: 175,35 g/mol
MDL: MFCD04972403
CAS-Nummer: 937595-71-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR