2-Chlor-5-fluor-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-5-fluoropyridine-3-boronic acid

BB-8376-1G BB-8376-250M
COBBBB-8376-1GEA 864 EUR
COBBBB-8376-1G COBBBB-8376-250M
2-Chlor-5-fluor-3-pyridinboronsäure
2-Chloro-5-fluoro-3-pyridineboronic acid
Formel: C₅H₄BClFNO₂
Molecular Weight: 175.35 g/mol
MDL: MFCD03094988
CAS-Nummer: 913373-43-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR