5-Chlor-2-methoxy-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
5-Chloro-2-methoxypyridine-3-boronic acid, (5-Chloro-2-methoxypyridin-3-yl)boronic acid
BB-8469-1G
BB-8469-250M
COBBBB-8469-1GEA
302
EUR
COBBBB-8469-1G
COBBBB-8469-250M
5-Chlor-2-methoxy-3-pyridinboronsäure
5-Chloro-2-methoxy-3-pyridineboronic acid
Formel:
C₆H₇BClNO₃ Molecular Weight: 187,39 g/mol |
MDL:
MFCD04114579 CAS-Nummer: 943153-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...