2-Chlor-5-fluor-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-5-fluoropyridine-4-boronic Acid

BB-8474-1G BB-8474-250M
COBBBB-8474-1GEA 246 EUR
COBBBB-8474-1G COBBBB-8474-250M
2-Chlor-5-fluor-4-pyridinboronsäure
2-Chloro-5-fluoro-4-pyridineboronic acid
Formel: C₅H₄BClFNO₂
Molecular Weight: 175.35 g/mol
MDL: MFCD04972404
CAS-Nummer: 951677-47-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR