2,5-Dichlor-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2,5-Dichloropyridine-3-boronic acid
BB-8499-1G
BB-8499-250M
COBBBB-8499-1GEA
312
EUR
COBBBB-8499-1G
COBBBB-8499-250M
2,5-Dichlor-3-pyridinboronsäure
2,5-Dichloro-3-pyridineboronic acid
Formel:
C₅H₄BCl₂NO₂ Molecular Weight: 191,81 g/mol |
MDL:
MFCD06659486 CAS-Nummer: 536693-97-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...