2-Brom-5-chlor-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-5-chloropyridine-4-boronic acid, (2-Bromo-5-chloropyridin-4-yl)boronic acid

BB-8542-5G BB-8542-1G
COBBBB-8542-5GEA 1180 EUR
COBBBB-8542-5G COBBBB-8542-1G
2-Brom-5-chlor-4-pyridinboronsäure
2-Bromo-5-chloro-4-pyridineboronic acid
Formel: C₅H₄BBrClNO₂
Molecular Weight: 236.26 g/mol
MDL: MFCD08064041
CAS-Nummer: 1072952-51-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR