4-Methoxypicolinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Methoxypyridine-2-carboxylic acid

CA-4030-5G CA-4030-1G
COBBCA-4030-5GEA 191 EUR
COBBCA-4030-5G COBBCA-4030-1G
4-Methoxypicolinsäure
4-Methoxypicolinsäure
Formel: C₇H₇NO₃
Molecular Weight: 153,14 g/mol
MDL: MFCD04115716
CAS-Nummer: 29082-91-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR