4-Chlorpicolinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloropyridine-2-carboxylic acid

CA-4035-5G CA-4035-25G CA-4035-1G CA-4035-100G
COBBCA-4035-5GEA 25.9 EUR
COBBCA-4035-5G COBBCA-4035-25G COBBCA-4035-1G COBBCA-4035-100G
4-Chlorpicolinsäure
4-Chlorpicolinsäure
Formel: C₆H₄ClNO₂
Molecular Weight: 157,56 g/mol
MDL: MFCD00191400
CAS-Nummer: 5470-22-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR