4-Brompicolinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromopyridine-2-carboxylic acid

CA-4037-1G CA-4037-5G
COBBCA-4037-1GEA 25.9 EUR
COBBCA-4037-1G COBBCA-4037-5G
4-Brompicolinsäure
4-Brompicolinsäure
Formel: C₆H₄BrNO₂
Molecular Weight: 202.01 g/mol
MDL: MFCD03426665
CAS-Nummer: 30766-03-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR