3-Fluorisonicotinsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Fluoropyridine-4-carboxylic acid
CA-4071-5G
CA-4071-25G
CA-4071-1G
COBBCA-4071-5GEA
51.3
EUR
COBBCA-4071-5G
COBBCA-4071-25G
COBBCA-4071-1G
3-Fluorisonicotinsäure
3-Fluorisonicotinsäure
Formel:
C₆H₄FNO₂ Molecular Weight: 141,1 g/mol |
MDL:
MFCD03789251 CAS-Nummer: 393-53-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...