Methyl-6-aminonicotinat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 6-aminonicotinate, 6-Aminonicotinic acid methyl ester
CA-4102-5G
CA-4102-25G
CA-4102-1G
COBBCA-4102-5GEA
51
EUR
COBBCA-4102-5G
COBBCA-4102-25G
COBBCA-4102-1G
Methyl-6-aminonicotinat
Methyl-6-aminonicotinat
Formel:
C₇H₈N₂O₂ Molecular Weight: 152.15 g/mol |
MDL:
MFCD00797844 CAS-Nummer: 36052-24-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...