Methyl-6-hydroxy-5-nitronicotinat

Lieferant: COMBI-BLOCKS

CA-4121-5G CA-4121-1G
COBBCA-4121-5GEA 576 EUR
COBBCA-4121-5G COBBCA-4121-1G
Methyl-6-hydroxy-5-nitronicotinat
Methyl-6-hydroxy-5-nitronicotinat
Formel: C₇H₆N₂O₅
MDL: MFCD10699641
CAS-Nummer: 222970-61-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR