4-Iodpicolinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Iodopyridine-2-carboxylic acid

CA-4129-5G CA-4129-1G
COBBCA-4129-5GEA 0 EUR
COBBCA-4129-5G COBBCA-4129-1G
4-Iodpicolinsäure
4-Iodpicolinsäure
Formel: C₆H₄INO₂
MDL: MFCD03426666
CAS-Nummer: 405939-79-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR