Methyl-2-hydroxy-5-nitronicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 5-nitro-2-oxo-1,2-dihydropyridine-3-carboxylate

CA-4134-1G CA-4134-5G
COBBCA-4134-1GEA 141 EUR
COBBCA-4134-1G COBBCA-4134-5G
Methyl-2-hydroxy-5-nitronicotinat
Methyl-2-hydroxy-5-nitronicotinat
Formel: C₇H₆N₂O₅
Molecular Weight: 198,14 g/mol
MDL: MFCD07783889
CAS-Nummer: 856579-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR