6-Brompicolinsäure
Lieferant: COMBI-BLOCKS
Synonyme:
6-Bromo-2-pyridinecarboxylic acid, 6-Bromopyridine-2-carboxylic acid
CA-4148-5G
CA-4148-25G
CA-4148-100G
COBBCA-4148-5GEA
49.3
EUR
COBBCA-4148-5G
COBBCA-4148-25G
COBBCA-4148-100G
6-Brompicolinsäure
6-Brompicolinsäure
Formel:
C₆H₄BrNO₂ Molecular Weight: 202,01 g/mol |
MDL:
MFCD00093197 CAS-Nummer: 21190-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...