Methyl-6-hydroxynicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 6-oxo-1,6-dihydro-3-pyridinecarboxylate, Methyl 6-hydroxynicotinate

CA-4213-5G CA-4213-25G CA-4213-1G
COBBCA-4213-5GEA 96.7 EUR
COBBCA-4213-5G COBBCA-4213-25G COBBCA-4213-1G
Methyl-6-hydroxynicotinat
Methyl-6-hydroxynicotinat
Formel: C₇H₇NO₃
Molecular Weight: 153,14 g/mol
MDL: MFCD04113616
CAS-Nummer: 66171-50-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR