Methyl-6-chlornicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 6-chloronicotinate

CA-4214-25G CA-4214-5G CA-4214-100G
COBBCA-4214-25GEA 63.4 EUR
COBBCA-4214-25G COBBCA-4214-5G COBBCA-4214-100G
Methyl-6-chlornicotinat
Methyl-6-chlornicotinat
Formel: C₇H₆ClNO₂
Molecular Weight: 171,58 g/mol
MDL: MFCD00023420
CAS-Nummer: 73781-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR