Methyl-6-methoxynicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 6-methoxynicotinate, 6-Methoxynicotinic acid methylester, Methyl 2-methoxy-5-pyridinecarboxylate

CA-4215-5G CA-4215-25G
COBBCA-4215-5GEA 51.3 EUR
COBBCA-4215-5G COBBCA-4215-25G
Methyl-6-methoxynicotinat
Methyl-6-methoxynicotinat
Formel: C₈H₉NO₃
Molecular Weight: 167,16 g/mol
MDL: MFCD00674130
CAS-Nummer: 26218-80-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR