5-Brom-2-pyridincarbonitril

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromopicolinonitrile, 5-Bromo-2-cyanopyridine

CA-4305-5G CA-4305-25G CA-4305-100G
COBBCA-4305-5GEA 35.5 EUR
COBBCA-4305-5G COBBCA-4305-25G COBBCA-4305-100G
5-Brom-2-pyridincarbonitril
5-Brom-2-pyridincarbonitril
Formel: C₆H₃BrN₂
Molecular Weight: 183,01 g/mol
MDL: MFCD00234144
CAS-Nummer: 97483-77-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR