3-Aminoisonicotinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Aminoisonicotinic Acid, 3-Aminopyridine-4-carboxylic acid

CA-4309-1G CA-4309-5G CA-4309-25G
COBBCA-4309-1GEA 25.5 EUR
COBBCA-4309-1G COBBCA-4309-5G COBBCA-4309-25G
3-Aminoisonicotinsäure
3-Aminoisonicotinsäure
Formel: C₆H₆N₂O₂
Molecular Weight: 138,13 g/mol
MDL: MFCD00137840
CAS-Nummer: 7579-20-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR