2,6-Dichlor-5-fluornicotinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2,6-Dichloro-5-fluoropyridine-3-carboxylic acid

OS-7532-5G OS-7532-100G OS-7532-25G
COBBOS-7532-5GEA 26.2 EUR
COBBOS-7532-5G COBBOS-7532-100G COBBOS-7532-25G
2,6-Dichlor-5-fluornicotinsäure
2,6-Dichlor-5-fluornicotinsäure
Formel: C₆H₂Cl₂FNO₂
Molecular Weight: 209,99 g/mol
MDL: MFCD00799517
CAS-Nummer: 82671-06-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR