5-Fluorpicolinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Fluoro-2-pyridinecarboxylic acid, 5-Fluoro-2-picolinic acid, 5-Fluoropyridine-2-carboxylic acid

OS-7580-5G OS-7580-1G
COBBOS-7580-5GEA 99.6 EUR
COBBOS-7580-5G COBBOS-7580-1G
5-Fluorpicolinsäure
5-Fluorpicolinsäure
Formel: C₆H₄FNO₂
Molecular Weight: 141,1 g/mol
MDL: MFCD04114196
CAS-Nummer: 107504-08-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR