Ethylpicolinat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 2-pyridinecarboxylate, Ethyl 2-picolinate

OS-7623-25G OS-7623-5G OS-7623-100G
COBBOS-7623-25GEA 51.3 EUR
COBBOS-7623-25G COBBOS-7623-5G COBBOS-7623-100G
Ethylpicolinat
Ethylpicolinat
Formel: C₈H₉NO₂
Molecular Weight: 151,16 g/mol
MDL: MFCD00006292
CAS-Nummer: 2524-52-9
EINECS: 219-758-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR