4-Bromisatosäureanhydrid

Lieferant: COMBI-BLOCKS

Synonyme: 7-Bromo-1H-benzo[d][1,3]oxazine-2,4-dione

OS-7633-1G OS-7633-5G
COBBOS-7633-1GEA 182 EUR
COBBOS-7633-1G COBBOS-7633-5G
4-Bromisatosäureanhydrid
4-Bromisatosäureanhydrid
Formel: C₈H₄BrNO₃
Molecular Weight: 242,03 g/mol
MDL: MFCD11113115
CAS-Nummer: 76561-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR