1-Propyl-4-ethinylbenzol

Lieferant: COMBI-BLOCKS

Synonyme: (4-Propylphenyl)ethin, 4-Propyl-1-ethinylbenzol, 1-Ethinyl-4-propylbenzol, 4-Ethinyl-1-propylbenzol, (4-Propylphenyl)acetylen

OT-0594-5G OT-0594-25G OT-0594-1G
COBBOT-0594-5GEA 46.2 EUR
COBBOT-0594-5G COBBOT-0594-25G COBBOT-0594-1G
1-Propyl-4-ethinylbenzol
1-Propyl-4-ethinylbenzol
Formel: C₁₁H₁₂
Molecular Weight: 144,22 g/mol
MDL: MFCD00173886
CAS-Nummer: 62452-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR