1-Pentyl-4-ethinylbenzol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Pentyl-1-ethinylbenzol, (4-Pentylphenyl)acetylen, 1-Ethinyl-4-pentylbenzol, 4-Ethinyl-1-pentylbenzol, (4-Pentylphenyl)ethin

OT-0595-25G OT-0595-5G OT-0595-100G
COBBOT-0595-25GEA 154 EUR
COBBOT-0595-25G COBBOT-0595-5G COBBOT-0595-100G
1-Pentyl-4-ethinylbenzol
1-Pentyl-4-ethinylbenzol
Formel: C₁₃H₁₆
Molecular Weight: 172.27 g/mol
MDL: MFCD00173884
CAS-Nummer: 79887-10-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR