2,5-Dichlorisonicotinsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2,5-Dichloropyridine-4-carboxylic acid
PY-1186-5G
PY-1186-25G
PY-1186-1G
COBBPY-1186-5GEA
63.4
EUR
COBBPY-1186-5G
COBBPY-1186-25G
COBBPY-1186-1G
2,5-Dichlorisonicotinsäure
2,5-Dichlorisonicotinsäure
Formel:
C₆H₃Cl₂NO₂ Molecular Weight: 192 g/mol |
MDL:
MFCD00466640 CAS-Nummer: 88912-26-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...