3-Fluorpicolinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-fluoropyridine-2-carboxylic acid, 3-Fluoro-2-pyridinecarboxylic acid

PY-1315-5G PY-1315-1G
COBBPY-1315-5GEA 105 EUR
COBBPY-1315-5G COBBPY-1315-1G
3-Fluorpicolinsäure
3-Fluorpicolinsäure
Formel: C₆H₄FNO₂
Molecular Weight: 141,1 g/mol
MDL: MFCD04114108
CAS-Nummer: 152126-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR