5-Brom-2-nitropyridin

Lieferant: COMBI-BLOCKS

PY-7043-100G PY-7043-25G PY-7043-5G
COBBPY-7043-100GEA 141 EUR
COBBPY-7043-100G COBBPY-7043-25G COBBPY-7043-5G
5-Brom-2-nitropyridin
5-Brom-2-nitropyridin
Formel: C₅H₃BrN₂O₂
Molecular Weight: 203 g/mol
MDL: MFCD00160411
CAS-Nummer: 39856-50-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR