5-Chlor-2-hydroxy-3-nitropyridin
Lieferant: COMBI-BLOCKS
Synonyme:
5-Chloro-3-nitropyridin-2-ol
PY-7230-5G
PY-7230-1G
COBBPY-7230-5GEA
591
EUR
COBBPY-7230-5G
COBBPY-7230-1G
5-Chlor-2-hydroxy-3-nitropyridin
5-Chlor-2-hydroxy-3-nitropyridin
Formel:
C₅H₃ClN₂O₃ Molecular Weight: 174.54 g/mol |
MDL:
MFCD00114884 CAS-Nummer: 21427-61-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...