5-Chlor-2-ethoxy-3-nitropyridin

Lieferant: COMBI-BLOCKS

PY-7231-1G PY-7231-5G
COBBPY-7231-1GEA 124 EUR
COBBPY-7231-1G COBBPY-7231-5G
5-Chlor-2-ethoxy-3-nitropyridin
5-Chlor-2-ethoxy-3-nitropyridin
Formel: C₇H₇ClN₂O₃
MDL: MFCD07375033
CAS-Nummer: 886373-32-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR