Methyl-4-methoxynicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-Methoxynicotinate

PY-7253-1G PY-7253-5G
COBBPY-7253-1GEA 251 EUR
COBBPY-7253-1G COBBPY-7253-5G
Methyl-4-methoxynicotinat
Methyl-4-methoxynicotinat
Formel: C₈H₉NO₃
Molecular Weight: 167,16 g/mol
MDL: MFCD09864940
CAS-Nummer: 10177-32-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR