3-Methylpicolinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Methylpyridine-2-carboxylic acid

PY-7283-1G PY-7283-25G PY-7283-5G
COBBPY-7283-1GEA 26.2 EUR
COBBPY-7283-1G COBBPY-7283-25G COBBPY-7283-5G
3-Methylpicolinsäure
3-Methylpicolinsäure
Formel: C₇H₇NO₂
Molecular Weight: 137.14 g/mol
MDL: MFCD00191553
CAS-Nummer: 4021-07-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR