Methyl-4-hydroxynicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-hydroxynicotinate

PY-7325-1G PY-7325-250M
COBBPY-7325-1GEA 742 EUR
COBBPY-7325-1G COBBPY-7325-250M
Methyl-4-hydroxynicotinat
Methyl-4-hydroxynicotinat
Formel: C₇H₇NO₃
Molecular Weight: 153,14 g/mol
MDL: MFCD04114270
CAS-Nummer: 67367-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR