Methyl-6-(dimethylamino)nicotinat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 6-dimethylamino-3-pyridinecarboxylate
PY-7398-1G
PY-7398-5G
COBBPY-7398-1GEA
204
EUR
COBBPY-7398-1G
COBBPY-7398-5G
Methyl-6-(dimethylamino)nicotinat
Methyl-6-(dimethylamino)nicotinat
Formel:
C₉H₁₂N₂O₂ Molecular Weight: 180,21 g/mol |
MDL:
MFCD10699718 CAS-Nummer: 26218-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...