Ethyl-2-chlor-5-nitronicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 2-chloro-5-nitronicotinate

PY-7739-1G PY-7739-250M
COBBPY-7739-1GEA 313 EUR
COBBPY-7739-1G COBBPY-7739-250M
Ethyl-2-chlor-5-nitronicotinat
Ethyl-2-chlor-5-nitronicotinat
Formel: C₈H₇ClN₂O₄
Molecular Weight: 230.61 g/mol
MDL: MFCD07368896
CAS-Nummer: 151322-83-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR