2,5-Dichlor-3-nitropyridin

Lieferant: COMBI-BLOCKS

PY-7762-25G PY-7762-5G
COBBPY-7762-25GEA 130 EUR
COBBPY-7762-25G COBBPY-7762-5G
2,5-Dichlor-3-nitropyridin
2,5-Dichloro-3-nitropyridine
Formel: C₅H₂Cl₂N₂O₂
MDL: MFCD06658963
CAS-Nummer: 21427-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR