Methyl-2-aminonicotinat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 2-aminonicotinate
PY-7811-1G
PY-7811-25G
PY-7811-5G
COBBPY-7811-1GEA
26.2
EUR
COBBPY-7811-1G
COBBPY-7811-25G
COBBPY-7811-5G
Methyl-2-aminonicotinat
Methyl-2-aminonicotinat
Formel:
C₇H₈N₂O₂ Molecular Weight: 152.15 g/mol |
MDL:
MFCD00128872 CAS-Nummer: 14667-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...